Gene Transcripts

Gene Symbol
KRTAP4-6
Entrez Gene ID
81871
Gene Name
keratin associated protein 4-6
Gene Aliases
KAP4.15, KAP4.6, KRTAP4-15, KRTAP4.15
Chromosome Location
Chr. 17: 41139433 - 41140487 on Build GRCh38
UniGene ID
Hs.307014
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030976.1 NP_112238.1 1 852
GenBank mRNA AJ406938.1 - 1 551
AJ406945.1 - 1 816

View Full Product Details