Gene Transcripts
- Gene Symbol
- FAM83B
- Entrez Gene ID
- 222584
- Gene Name
- family with sequence similarity 83 member B
- Gene Aliases
- C6orf143
- Chromosome Location
- Chr. 6: 54846323 - 54945099 on Build GRCh38
- UniGene ID
- Hs.152423
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BG743165.1 |
- |
1 |
655 |
|
Chr. 6 - 54943952 - 54944665 [+] |
GenBankmRNA |
AA773657.1 |
- |
1 |
242 |
|
Chr. 6 - 54944366 - 54944661 [+] |
GenBankmRNA |
BM786734.1 |
- |
1 |
273 |
|
Chr. 6 - 54944335 - 54944998 [+] |
View Full Product Details