Gene Transcripts

Gene Symbol
FAM83B
Entrez Gene ID
222584
Gene Name
family with sequence similarity 83 member B
Gene Aliases
C6orf143
Chromosome Location
Chr. 6: 54846323 - 54945099 on Build GRCh38
UniGene ID
Hs.152423
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001010872.2 NP_001010872.1 5 4944
GenBank mRNA AK024927.1 - 1 430

View Full Product Details