Gene Transcripts
- Gene Symbol
- FRMD4A
- Entrez Gene ID
- 55691
- Gene Name
- FERM domain containing 4A
- Gene Aliases
- CCAFCA, FRMD4, bA295P9.4
- Chromosome Location
- Chr. 10: 13643706 - 14330952 on Build GRCh38
- UniGene ID
- Hs.330463
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK093978.1 |
- |
1 |
1707 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
EL955225.1 |
- |
1 |
272 |
|
Chr. 10 - 13689966 - 13690352 [-] |
View Full Product Details