Gene Transcripts

Gene Symbol
RSF1
Entrez Gene ID
51773
Gene Name
remodeling and spacing factor 1
Gene Aliases
HBXAP, RSF-1, XAP8, p325
Chromosome Location
Chr. 11: 77660009 - 77872232 on Build GRCh38
UniGene ID
Hs.420229
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_005274051.2 XP_005274108.1 16 6615
XM_017017923.1 XP_016873412.1 16 6363
GenBank mRNA AK096617.1 - 1 185

View Full Product Details