Gene Transcripts

Gene Symbol
CEP41
Entrez Gene ID
95681
Gene Name
centrosomal protein 41
Gene Aliases
JBTS15, TSGA14
Chromosome Location
Chr. 7: 130393771 - 130441210 on Build GRCh38
UniGene ID
Hs.368315
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001257160.1 NP_001244089.1 3 2905
GenBank mRNA AF429309.1 - 3 2751
AK001820.1 - 1 1650

View Full Product Details