Gene Transcripts
- Gene Symbol
- COG5
- Entrez Gene ID
- 10466
- Gene Name
- component of oligomeric golgi complex 5
- Gene Aliases
- CDG2I, GOLTC1, GTC90
- Chromosome Location
- Chr. 7: 107201743 - 107564514 on Build GRCh38
- UniGene ID
- Hs.239631
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK123726.1 |
- |
1 |
732 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CN284531.1 |
- |
1 |
41 |
|
Chr. 7 - 107203651 - 107204265 [-] |
View Full Product Details