Gene Transcripts
- Gene Symbol
- Palld
- Entrez Gene ID
- 72333
- Gene Name
- palladin, cytoskeletal associated protein
- Gene Aliases
- 2410003B16Rik, 6030492A02
- Chromosome Location
- Chr. 8: 61511429 - 61902700 on Build GRCm38
- UniGene ID
- Mm.29933
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK076423.1 |
- |
19 |
3672 |
|
AK077756.1 |
- |
1 |
338 |
|
BC127081.1 |
- |
13 |
2806 |
|
View Full Product Details