Gene Transcripts
- Gene Symbol
- Sgk3
- Entrez Gene ID
- 170755
- Gene Name
- serum/glucocorticoid regulated kinase 3
- Gene Aliases
- 2510015P22Rik, A330005P07Rik, Cisk, fy, fz
- Chromosome Location
- Chr. 1: 9798130 - 9902570 on Build GRCm38
- UniGene ID
- Mm.336410
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK080469.1 |
- |
1 |
3146 |
|
View Full Product Details