Gene Transcripts
- Gene Symbol
- Pcnt
- Entrez Gene ID
- 18541
- Gene Name
- pericentrin (kendrin)
- Gene Aliases
- AW476095, C86676, KEN, Pcnt2, kendrin, m239Asp, m275Asp
- Chromosome Location
- Chr. 10: 76351254 - 76444122 on Build GRCm38
- UniGene ID
- Mm.251794
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK052041.1 |
- |
1 |
1550 |
|
View Full Product Details