Gene Transcripts
- Gene Symbol
- Igdcc3
- Entrez Gene ID
- 19289
- Gene Name
- immunoglobulin superfamily, DCC subclass, member 3
- Gene Aliases
- 2810401C09Rik, AI851425, Punc, WI-14920
- Chromosome Location
- Chr. 9: 65141121 - 65187031 on Build GRCm38
- UniGene ID
- Mm.392333
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK012957.1 |
- |
1 |
38 |
|
View Full Product Details