Gene Transcripts
- Gene Symbol
- FALEC
- Entrez Gene ID
- 100874054
- Gene Name
- focally amplified long non-coding RNA in epithelial cancer
- Gene Aliases
- FAL1, LINC00568, ncRNA-a1
- Chromosome Location
- Chr. 1: 150515757 - 150518032 on Build GRCh38
- UniGene ID
- Hs.576460
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_051960.1 |
|
2 |
507 |
|
GenBank mRNA |
AL713297.1 |
- |
2 |
508 |
|
View Full Product Details