Gene Transcripts
- Gene Symbol
- COX10
- Entrez Gene ID
- 1352
- Gene Name
- COX10, heme A:farnesyltransferase cytochrome c oxidase assembly factor
- Gene Aliases
- Chromosome Location
- Chr. 17: 14069402 - 14208679 on Build GRCh38
- UniGene ID
- Hs.462278
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC017752.1 |
- |
3 |
932 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000458492.1 |
- |
3 |
932 |
|
Chr. 17 - 14210488 - 14217922 [+] |
View Full Product Details