Gene Transcripts
- Gene Symbol
- BOK-AS1
- Entrez Gene ID
- 100379249
- Gene Name
- BOK antisense RNA 1
- Gene Aliases
- BOK-AS, BOKAS, NAToB, NCRNA00151
- Chromosome Location
- Chr. 2: 241544384 - 241559143 on Build GRCh38
- UniGene ID
- Hs.621128
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_033346.1 |
|
2 |
516 |
|
GenBank mRNA |
FJ185168.1 |
- |
2 |
516 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000434306.1 |
- |
2 |
350 |
|
Chr. 2 - 241544403 - 241558977 [-] |
View Full Product Details