Gene Transcripts
- Gene Symbol
- DSE
- Entrez Gene ID
- 29940
- Gene Name
- dermatan sulfate epimerase
- Gene Aliases
- DS-epi1, DSEP, DSEPI, EDSMC2, SART-2, SART2
- Chromosome Location
- Chr. 6: 116254152 - 116441261 on Build GRCh38
- UniGene ID
- Hs.458358
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_136520.1 |
|
2 |
896 |
|
GenBank mRNA |
AK093256.1 |
- |
2 |
775 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000448740.2 |
- |
2 |
841 |
|
Chr. 6 - 116254207 - 116256743 [+] |
View Full Product Details