Gene Transcripts
- Gene Symbol
- MLK7-AS1
- Entrez Gene ID
- 339751
- Gene Name
- MLK7 antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 2: 173197712 - 173282036 on Build GRCh38
- UniGene ID
- Hs.570083
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_033882.1 |
|
8 |
1702 |
|
GenBank mRNA |
BC040565.1 |
- |
8 |
1712 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000423106.2 |
- |
8 |
1702 |
|
Chr. 2 - 173197712 - 173282036 [-] |
View Full Product Details