Gene Transcripts
- Gene Symbol
- ENSG00000240050
- Entrez Gene ID
- ENSG00000240050
- Gene Name
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
- Gene Symbol
- DSE
- Entrez Gene ID
- 29940
- Gene Name
- dermatan sulfate epimerase
- Gene Aliases
- DS-epi1, DSEP, DSEPI, EDSMC2, SART-2, SART2
- Chromosome Location
- Chr. 6: 116254152 - 116441261 on Build GRCh38
- UniGene ID
- Hs.458358
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DC416992.1 |
- |
2 |
241 |
|
Chr. 6 - 116280086 - 116399276 [+] |
View Full Product Details