Gene Transcripts
- Gene Symbol
- UGDH-AS1
- Entrez Gene ID
- 100885776
- Gene Name
- UGDH antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 4: 39527839 - 39594707 on Build GRCh38
- UniGene ID
- Hs.640769
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_047679.1 |
|
3, 4 |
792 |
|
GenBank mRNA |
AK056558.1 |
- |
3, 4 |
612 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000504032.1 |
- |
3, 4 |
612 |
|
Chr. 4 - 39528019 - 39594707 [+] |
View Full Product Details