Gene Transcripts

Gene Symbol
Suox
Entrez Gene ID
211389
Gene Name
sulfite oxidase
Gene Aliases
SO
Chromosome Location
Chr. 10: 128669887 - 128673918 on Build GRCm38
UniGene ID
Mm.23352
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173733.3 NP_776094.2 4 337
GenBank mRNA AK149422.1 - 4 325
AK154608.1 - 4 325
BC027197.1 - 3 292 4160277

View Full Product Details