Gene Transcripts

Gene Symbol
Plekhh1
Entrez Gene ID
211945
Gene Name
pleckstrin homology domain containing, family H (with MyTH4 domain) member 1
Gene Aliases
AV308913, C77772, D630024D12Rik, mKIAA1200
Chromosome Location
Chr. 12: 79029163 - 79081655 on Build GRCm38
UniGene ID
Mm.478321
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181073.3 NP_851418.2 12 2109
GenBank mRNA AK122464.1 - 12 2078
AK143387.1 - 12 1926

View Full Product Details