Gene Transcripts

Gene Symbol
Sertad4
Entrez Gene ID
214791
Gene Name
SERTA domain containing 4
Gene Aliases
4833404A09, C130018M11Rik
Chromosome Location
Chr. 1: 192844469 - 192855752 on Build GRCm38
UniGene ID
Mm.22806
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001177794.1 NP_001171265.1 4 635
NM_198247.2 NP_937890.2 4 890
GenBank mRNA BC060251.1 - 4 859

View Full Product Details