Gene Transcripts

Gene Symbol
Trim58
Entrez Gene ID
216781
Gene Name
tripartite motif-containing 58
Gene Aliases
Gm16, Gm1783
Chromosome Location
Chr. 11: 58640465 - 58652404 on Build GRCm38
UniGene ID
Mm.339507
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039047.1 NP_001034136.1 2 450
GenBank mRNA BC147805.1 - 2 469
BC147806.1 - 2 469 9087523
BC150936.1 - 2 524 9087847

View Full Product Details