Gene Transcripts
- Gene Symbol
- Myo15b
- Entrez Gene ID
- 217328
- Gene Name
- myosin XVB
- Gene Aliases
- E330039G21Rik, Gm688, Gm886, mKIAA1783
- Chromosome Location
- Chr. 11: 115858391 - 115892594 on Build GRCm38
- UniGene ID
- Mm.308940
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK087906.2 |
- |
1 |
94 |
|
AK173262.1 |
- |
21 |
2367 |
|
View Full Product Details