Gene Transcripts

Gene Symbol
SERPINB5
Entrez Gene ID
5268
Gene Name
serpin family B member 5
Gene Aliases
PI5, maspin
Chromosome Location
Chr. 18: 63476911 - 63505085 on Build GRCh38
UniGene ID
Hs.55279
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK312765.1 - 4 445
BC020713.1 - 4 420 4248249
U04313.1 - 4 400

View Full Product Details