Gene Transcripts

Gene Symbol
Ak5
Entrez Gene ID
229949
Gene Name
adenylate kinase 5
Gene Aliases
Chromosome Location
Chr. 3: 152462815 - 152668376 on Build GRCm38
UniGene ID
Mm.207057
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081277.1 NP_001074746.1 7 1205
GenBank mRNA AK053807.1 - 7 1205

View Full Product Details