Gene Transcripts

Gene Symbol
Mb21d2
Entrez Gene ID
239796
Gene Name
Mab-21 domain containing 2
Gene Aliases
1600021P15Rik, A430031N04, C87006
Chromosome Location
Chr. 16: 28826176 - 28929698 on Build GRCm38
UniGene ID
Mm.155887
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177718.3 NP_808386.2 2 260
GenBank mRNA AK079722.1 - 3 220
BC068245.1 - 2 248
BC085092.1 - 2 186 30735763
BE983254.1 - 2 260

View Full Product Details