Gene Transcripts

Gene Symbol
Oas3
Entrez Gene ID
246727
Gene Name
2'-5' oligoadenylate synthetase 3
Gene Aliases
Oasl10
Chromosome Location
Chr. 5: 120753098 - 120777659 on Build GRCm38
UniGene ID
Mm.204887
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145226.2 NP_660261.1 4 834
GenBank mRNA AB067534.1 - 4 834
AF453830.1 - 4 826
AK158392.1 - 4 816
BC141055.1 - 4 827 9055667

View Full Product Details