Gene Transcripts
- Gene Symbol
- Casc4
- Entrez Gene ID
- 319996
- Gene Name
- cancer susceptibility candidate 4
- Gene Aliases
- D130060C09Rik, VGFG2573
- Chromosome Location
- Chr. 2: 121866970 - 121936220 on Build GRCm38
- UniGene ID
- Mm.292109
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001205369.1 |
NP_001192298.1
|
4, 5 |
822 |
|
NM_001205370.1 |
NP_001192299.1
|
4, 5 |
822 |
|
NM_001205371.1 |
NP_001192300.1
|
4, 5 |
412 |
|
NM_177054.5 |
NP_796028.2
|
4, 5 |
822 |
|
NM_199038.3 |
NP_950239.2
|
4, 5 |
822 |
|
GenBank mRNA |
AK131751.1 |
- |
4, 5 |
413 |
|
AK160708.1 |
- |
4, 5 |
663 |
|
AY484583.1 |
- |
4, 5 |
662 |
|
AY484585.1 |
- |
4, 5 |
662 |
|
BC034556.1 |
- |
4, 5 |
685 |
|
BC064446.1 |
- |
4, 5 |
822 |
6843743 |
View Full Product Details