Gene Transcripts

Gene Symbol
SFRP4
Entrez Gene ID
6424
Gene Name
secreted frizzled related protein 4
Gene Aliases
FRP-4, FRPHE, sFRP-4
Chromosome Location
Chr. 7: 37905932 - 37916923 on Build GRCh38
UniGene ID
Hs.658169
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003014.3 NP_003005.2 5 1231
GenBank mRNA AF026692.1 - 5 1102
AK302357.1 - 5 1153
AK303977.1 - 6 1051
BC047684.1 - 5 1089 5742936
BC058911.1 - 5 1109 5228231
BT019679.1 - 5 845
CR541731.1 - 5 845
CR541755.1 - 5 845
DQ420628.1 - 5 864

View Full Product Details