Gene Transcripts

Gene Symbol
L1cam
Entrez Gene ID
50687
Gene Name
L1 cell adhesion molecule
Gene Aliases
Hsas, Hyd, NCAML1
Chromosome Location
Chr. X: 156911336 - 156928056 on Build Rnor_6.0
UniGene ID
Rn.10378
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017345.1 NP_059041.1 26, 27 3636
XM_008773637.1 XP_008771859.1 25, 26 3630
GenBank mRNA X59149.1 - 26, 27 3636

View Full Product Details