Gene Transcripts

Gene Symbol
Lmna
Entrez Gene ID
60374
Gene Name
lamin A/C
Gene Aliases
Chromosome Location
Chr. 2: 187842884 - 187863552 on Build Rnor_6.0
UniGene ID
Rn.44161
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002016.2 NP_001002016.2 6 1290
GenBank mRNA BC062018.1 - 5 845
CA338388.1 - 2 275
X66870.1 - 6 1249
X99257.1 - 6 749

View Full Product Details