Gene Transcripts

Gene Symbol
Nptx2
Entrez Gene ID
288475
Gene Name
neuronal pentraxin II
Gene Aliases
Narp
Chromosome Location
Chr. 12: 12014638 - 12025547 on Build Rnor_6.0
UniGene ID
Rn.162101
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034199.1 NP_001029371.1 5 1391
GenBank mRNA CO388671.1 - 1 139
S82649.1 - 1325

View Full Product Details