Gene Transcripts

Gene Symbol
Atp8b1
Entrez Gene ID
291555
Gene Name
ATPase, aminophospholipid transporter, class I, type 8B, member 1
Gene Aliases
Chromosome Location
Chr. 18: 60013388 - 60152836 on Build Rnor_6.0
UniGene ID
Rn.224747
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106140.1 NP_001099610.1 8, 9 782
XM_006254842.2 XP_006254904.1 8, 9 805

View Full Product Details