Gene Transcripts

Gene Symbol
Pcdhb7
Entrez Gene ID
291652
Gene Name
protocadherin beta 7
Gene Aliases
pcdh-T3
Chromosome Location
Chr. 18: 30423469 - 30428141 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_001055294.5 XP_001055294.3 1 2116

View Full Product Details