Gene Transcripts

Gene Symbol
Olr81
Entrez Gene ID
293224
Gene Name
olfactory receptor 81
Gene Aliases
Chromosome Location
Chr. 1: 168293741 - 168294679 on Build Rnor_6.0
UniGene ID
Rn.141905
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001271.1 NP_001001271.1 1 468

View Full Product Details