Gene Transcripts

Gene Symbol
Sfxn2
Entrez Gene ID
294011
Gene Name
sideroflexin 2
Gene Aliases
Chromosome Location
Chr. 1: 266333440 - 266345736 on Build Rnor_6.0
UniGene ID
Rn.230666
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013072.1 NP_001013090.1 2 158
GenBank mRNA BC081888.1 - 2 158

View Full Product Details