Gene Transcripts
- Gene Symbol
- Taar7g
- Entrez Gene ID
- 294131
- Gene Name
- trace amine-associated receptor 7g
- Gene Aliases
- Ta9
- Chromosome Location
- Chr. 1: 22465092 - 22466168 on Build Rnor_6.0
- UniGene ID
- Rn.138143
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_175588.1 |
NP_783178.1
|
1 |
1035 |
|
View Full Product Details