Gene Transcripts

Gene Symbol
Bsdc1
Entrez Gene ID
297890
Gene Name
BSD domain containing 1
Gene Aliases
RGD1311622
Chromosome Location
Chr. 5: 147661618 - 147689766 on Build Rnor_6.0
UniGene ID
Rn.3404
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106636.1 NP_001100106.1 9 748

View Full Product Details