Gene Transcripts
- Gene Symbol
- Rbm31y
- Entrez Gene ID
- 302780
- Gene Name
- RNA binding motif 31, Y-linked
- Gene Aliases
- RGD1562219
- Chromosome Location
- Chr. X: 98093734 - 98095719 on Build Rnor_6.0
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_229073.3 |
XP_229073.1
|
1 |
1660 |
|
View Full Product Details