Gene Transcripts

Gene Symbol
Rpgrip1l
Entrez Gene ID
307724
Gene Name
Rpgrip1-like
Gene Aliases
RGD1311099
Chromosome Location
Chr. 19: 17115412 - 17208055 on Build Rnor_6.0
UniGene ID
Rn.55934
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107414.1 NP_001100884.1 16 2291
XM_008772313.1 XP_008770535.1 15 2719

View Full Product Details