Gene Transcripts

Gene Symbol
HIST1H4B
Entrez Gene ID
8366
Gene Name
histone cluster 1, H4b
Gene Aliases
H4/I, H4FI
Chromosome Location
Chr. 6: 26026896 - 26027252 on Build GRCh38
UniGene ID
Hs.143080
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003544.2 NP_003535.1 1 66
GenBank mRNA BC069467.1 - 1 66

View Full Product Details