Gene Transcripts

Gene Symbol
Usp31
Entrez Gene ID
308959
Gene Name
ubiquitin specific peptidase 31
Gene Aliases
Chromosome Location
Chr. 1: 191615146 - 191651628 on Build Rnor_6.0
UniGene ID
Rn.64320
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107548.1 NP_001101018.1 15 2109

View Full Product Details