Gene Transcripts

Gene Symbol
Rdh12
Entrez Gene ID
314264
Gene Name
retinol dehydrogenase 12 (all-trans/9-cis/11-cis)
Gene Aliases
Chromosome Location
Chr. 6: 102392828 - 102405752 on Build Rnor_6.0
UniGene ID
Rn.148972
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108037.1 NP_001101507.1 5 683
XM_008764779.1 XP_008763001.1 4 813

View Full Product Details