Gene Transcripts

Gene Symbol
Fbxo43
Entrez Gene ID
315034
Gene Name
F-box protein 43
Gene Aliases
Chromosome Location
Chr. 7: 74925091 - 74938663 on Build Rnor_6.0
UniGene ID
Rn.137608
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012117.1 NP_001012117.2 2 759
XM_006241535.2 XP_006241597.1 2 517
GenBank mRNA BC082107.1 - 2 759

View Full Product Details