Gene Transcripts
- Gene Symbol
- ARSE
- Entrez Gene ID
- 415
- Gene Name
- arylsulfatase E (chondrodysplasia punctata 1)
- Gene Aliases
- ASE, CDPX, CDPX1, CDPXR
- Chromosome Location
- Chr. X: 2934632 - 2968310 on Build GRCh38
- UniGene ID
- Hs.386975
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000047.2 |
NP_000038.2
|
7, 8 |
1250 |
|
NM_001282628.1 |
NP_001269557.1
|
8, 9 |
1536 |
|
NM_001282631.1 |
NP_001269560.1
|
6, 7 |
1020 |
|
XM_005274518.2 |
XP_005274575.1
|
7, 8 |
1177 |
|
XM_005274519.4 |
XP_005274576.1
|
7, 8 |
1340 |
|
XM_005274521.4 |
XP_005274578.1
|
6, 7 |
1133 |
|
XM_017029525.1 |
XP_016885014.1
|
8, 9 |
1467 |
|
GenBank mRNA |
AK223183.1 |
- |
7, 8 |
1080 |
|
AK223199.1 |
- |
7, 8 |
1047 |
|
AK301004.1 |
- |
8, 9 |
1367 |
|
AK313093.1 |
- |
7, 8 |
1069 |
|
BC130438.1 |
- |
7, 8 |
1045 |
40146469 |
X83573.1 |
- |
7, 8 |
1067 |
|
View Full Product Details