Gene Transcripts

Gene Symbol
Map7d3
Entrez Gene ID
317614
Gene Name
MAP7 domain containing 3
Gene Aliases
RGD1565514
Chromosome Location
Chr. X: 159176825 - 159220494 on Build Rnor_6.0
UniGene ID
Rn.121716
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271327.1 NP_001258256.1 5 443
XM_008773649.1 XP_008771871.1 5 639
XM_008773650.1 XP_008771872.1 5 551

View Full Product Details