Gene Transcripts

Gene Symbol
Gpr25
Entrez Gene ID
363993
Gene Name
G protein-coupled receptor 25
Gene Aliases
Chromosome Location
Chr. 13: 53135815 - 53139179 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008769579.1 XP_008767801.1 1 2710

View Full Product Details