Gene Transcripts

Gene Symbol
Phox2b
Entrez Gene ID
364152
Gene Name
paired-like homeobox 2b
Gene Aliases
NBPhox
Chromosome Location
Chr. 14: 42711438 - 42718707 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008770167.1 XP_008768389.1 1, 2 3115

View Full Product Details