Gene Transcripts

Gene Symbol
Ankrd11
Entrez Gene ID
365023
Gene Name
ankyrin repeat domain 11
Gene Aliases
Chromosome Location
Chr. 19: 55703831 - 55795237 on Build Rnor_6.0
UniGene ID
Rn.23844
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008772666.1 XP_008770888.1 8 4696
XM_008772667.1 XP_008770889.1 8 4693
XM_008772668.1 XP_008770890.1 8 4687

View Full Product Details