Gene Transcripts

Gene Symbol
Umodl1
Entrez Gene ID
365544
Gene Name
uromodulin-like 1
Gene Aliases
RGD1566039
Chromosome Location
Chr. 20: 9643509 - 9703091 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008772853.1 XP_008771075.1 13 2578

View Full Product Details